siRNA Details
| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318049.1 | NP_001304978.1 | 3 | 1109 | ||
| NM_138969.3 | NP_620419.2 | 3 | 1109 | |||
| GenBank mRNA | AB083038.1 | - | 3 | 1015 | ||
| AK057667.1 | - | 3 | 451 | |||
| AK095159.1 | - | 3 | 1109 | |||
| BC037219.1 | - | 3 | 456 | 5181472 | ||
| BC064525.1 | - | 3 | 428 | 5752917 | ||

SDR16C5
Genomic Map