siRNA Details
| Gene Symbol: ZFC3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144982.4 | NP_659419.3 | 5 | 1810 | ||
| GenBank mRNA | AB011118.2 | - | 5 | 1660 | ||
| BX647709.1 | - | 5 | 1690 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFC3H1| Gene Symbol: ZFC3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144982.4 | NP_659419.3 | 5 | 1810 | ||
| GenBank mRNA | AB011118.2 | - | 5 | 1660 | ||
| BX647709.1 | - | 5 | 1690 | |||