siRNA Details
| Gene Symbol: MYRFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182530.2 | NP_872336.2 | 21, 22 | 2573 | ||
| XM_011539052.2 | XP_011537354.1 | 20, 21 | 3054 | |||
| XM_011539055.1 | XP_011537357.1 | 19, 20 | 2041 | |||
| XM_017018967.1 | XP_016874456.1 | 20, 21 | 2855 | |||
| XM_017018968.1 | XP_016874457.1 | 21, 22 | 3309 | |||
| XM_017018969.1 | XP_016874458.1 | 20, 21 | 2789 | |||
| XM_017018970.1 | XP_016874459.1 | 19, 20 | 2599 | |||
| XM_017018973.1 | XP_016874462.1 | 17, 18 | 2270 | |||
| GenBank mRNA | AK057785.1 | - | 8, 9 | 690 | ||
| BC101820.1 | - | 6, 7 | 525 | |||

MYRFL
Genomic Map