siRNA Details
Gene Symbol: FAM71C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153364.3 | NP_699195.1 | 1 | 984 | ||
GenBank mRNA | AK312767.1 | - | 1 | 980 | ||
BC031221.1 | - | 1 | 981 | 5272209 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM71C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153364.3 | NP_699195.1 | 1 | 984 | ||
GenBank mRNA | AK312767.1 | - | 1 | 980 | ||
BC031221.1 | - | 1 | 981 | 5272209 |