siRNA Details
| Gene Symbol: ARID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152641.2 | NP_689854.2 | 13, 14 | 1723 | ||
| XM_006719272.3 | XP_006719335.1 | 13, 14 | 1877 | |||
| XM_011538025.1 | XP_011536327.1 | 7, 8 | 830 | |||
| GenBank mRNA | AL832200.1 | - | 9, 10 | 1276 | ||
| AY727870.2 | - | 13, 14 | 1723 | |||
| BX648152.1 | - | 7, 8 | 1014 | |||
| BX649133.1 | - | 5, 6 | 569 | |||
| CR749833.1 | - | 12, 13 | 1642 | |||
| DQ096628.1 | - | 13, 14 | 1723 | |||

ARID2
Genomic Map