siRNA Details
| Gene Symbol: FAM227B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152647.2 | NP_689860.2 | 9 | 962 | ||
| XM_005254213.3 | XP_005254270.1 | 9 | 1039 | |||
| XM_005254214.3 | XP_005254271.1 | 9 | 972 | |||
| XM_005254215.3 | XP_005254272.1 | 8 | 1003 | |||
| XM_005254219.3 | XP_005254276.1 | 9 | 1039 | |||
| XM_005254220.3 | XP_005254277.1 | 9 | 1039 | |||
| XM_006720423.2 | XP_006720486.1 | 10 | 1004 | |||
| XM_006720424.2 | XP_006720487.1 | 9 | 1039 | |||
| XM_006720426.2 | XP_006720489.1 | 7 | 898 | |||
| XM_006720427.3 | XP_006720490.1 | 10 | 1010 | |||
| XM_011521319.2 | XP_011519621.1 | 9 | 2922 | |||
| XM_011521320.1 | XP_011519622.1 | 7 | 414 | |||
| XM_011521321.2 | XP_011519623.1 | 9 | 1039 | |||
| XM_011521322.1 | XP_011519624.1 | 6 | 378 | |||
| XM_011521324.1 | XP_011519626.1 | 9 | 1039 | |||
| XM_011521325.2 | XP_011519627.1 | 9 | 1039 | |||
| XM_017021991.1 | XP_016877480.1 | 10 | 1079 | |||
| XM_017021992.1 | XP_016877481.1 | 7 | 727 | |||
| XM_017021995.1 | XP_016877484.1 | 9 | 1039 | |||
| XM_017021996.1 | XP_016877485.1 | 8 | 487 | |||
| GenBank mRNA | AK057362.1 | - | 9 | 960 | ||

FAM227B
Genomic Map