siRNA Details
| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174916.2 | NP_777576.1 | 2 | 291 | ||
| GenBank mRNA | AF525401.1 | - | 2 | 174 | ||
| AK056441.1 | - | 2 | 241 | |||
| AK292516.1 | - | 2 | 238 | |||
| AK302489.1 | - | 2 | 318 | |||
| AY061886.1 | - | 2 | 225 | |||
| BC113505.1 | - | 2 | 303 | 8322557 | ||
| BC113507.1 | - | 2 | 303 | 8322559 | ||
 Back To Top
 Back To Top
 UBR1
UBR1 Genomic Map
Genomic Map Back To Top
 Back To Top