siRNA Details
| Gene Symbol: ZNF778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201407.1 | NP_001188336.1 | 4 | 528 | ||
| NM_182531.3 | NP_872337.2 | 4 | 528 | |||
| XM_005256288.2 | XP_005256345.1 | 4 | 514 | |||
| XM_011522940.2 | XP_011521242.1 | 4 | 481 | |||
| XM_017023015.1 | XP_016878504.1 | 3 | 436 | |||
| XM_017023016.1 | XP_016878505.1 | 4 | 507 | |||
| XM_017023017.1 | XP_016878506.1 | 4 | 507 | |||
| GenBank mRNA | AK056437.1 | - | 4 | 521 | ||
| BC125192.1 | - | 2 | 149 | 40122711 | ||
| DA297948.1 | - | 4 | 528 | |||
| DR001516.1 | - | 4 | 516 | |||

ZNF778
Genomic Map