siRNA Details
| Gene Symbol: WDR90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145294.4 | NP_660337.3 | 39 | 5029 | ||
| XM_017023023.1 | XP_016878512.1 | 40 | 5299 | |||
| XM_017023024.1 | XP_016878513.1 | 22 | 2912 | |||
| GenBank mRNA | AK092719.1 | - | 10 | 1745 | ||
| AK093609.1 | - | 4 | 1429 | |||
| AK131510.1 | - | 22 | 3335 | |||
| BC065836.1 | - | 4 | 941 | |||

WDR90
Genomic Map