siRNA Details
| Gene Symbol: WDR90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145294.4 | NP_660337.3 | 5 | 598 | ||
| XM_017023023.1 | XP_016878512.1 | 5 | 598 | |||
| GenBank mRNA | AB067511.1 | - | 4 | 986 | ||
| AK091062.1 | - | 5 | 598 | |||
| BC121185.1 | - | 5 | 608 | |||
| BC121186.1 | - | 5 | 608 | |||

WDR90
Genomic Map