siRNA Details
| Gene Symbol: ZNF627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290083.1 | NP_001277012.1 | 5 | 1651 | ||
| NM_001290084.1 | NP_001277013.1 | 4 | 1485 | |||
| NM_001290085.1 | NP_001277014.1 | 4 | 1492 | |||
| NM_145295.3 | NP_660338.1 | 4 | 1496 | |||
| XM_011527780.2 | XP_011526082.1 | 4 | 1384 | |||
| GenBank mRNA | AK074846.1 | - | 4 | 1404 | ||
| AK075063.1 | - | 1 | 584 | |||
| AK095639.1 | - | 4 | 1422 | |||
| BC006279.2 | - | 4 | 1417 | |||
| BC069232.1 | - | 4 | 1381 | 6645413 | ||
| BC098416.1 | - | 4 | 1467 | |||

ZNF627
Genomic Map