siRNA Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152658.2 | NP_689871.1 | 4 | 1669 | ||
| XM_005258623.2 | XP_005258680.1 | 4 | 1478 | |||
| XM_011526604.2 | XP_011524906.1 | 4 | 1678 | |||
| GenBank mRNA | AK057453.1 | - | 5 | 1522 | ||
| AK093048.1 | - | 4 | 1670 | |||
| AK296640.1 | - | 4 | 1163 | |||
| BC046207.1 | - | 4 | 1218 | 6176484 | ||
| BC072416.1 | - | 4 | 1309 | 6169558 | ||
| JQ410992.1 | - | 4 | 1546 | |||

THAP8
Genomic Map