siRNA Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152658.2 | NP_689871.1 | 4 | 1650 | ||
| XM_005258623.2 | XP_005258680.1 | 4 | 1459 | |||
| XM_011526604.2 | XP_011524906.1 | 4 | 1659 | |||
| GenBank mRNA | AK057453.1 | - | 5 | 1503 | ||
| AK093048.1 | - | 4 | 1651 | |||
| AK296640.1 | - | 4 | 1144 | |||
| AK297630.1 | - | 3 | 960 | |||
| BC046207.1 | - | 4 | 1199 | 6176484 | ||
| BC072416.1 | - | 4 | 1290 | 6169558 | ||
| JQ410992.1 | - | 4 | 1527 | |||

THAP8
Genomic Map