siRNA Details
| Gene Symbol: ALG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305242.1 | NP_001292171.1 | 2 | 246 | ||
| NM_144988.3 | NP_659425.1 | 2 | 246 | |||
| XM_005270582.3 | XP_005270639.1 | 2 | 268 | |||
| XM_011540897.2 | XP_011539199.1 | 2 | 268 | |||
| GenBank mRNA | AK289395.1 | - | 2 | 246 | ||
| AW242346.1 | - | 2 | 160 | |||
| BC011706.2 | - | 2 | 222 | 3689162 | ||

ALG14
Genomic Map