siRNA Details
| Gene Symbol: TMEM201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010866.3 | NP_001010866.1 | 3 | 330 | ||
| NM_001130924.2 | NP_001124396.2 | 3 | 330 | |||
| XM_006710417.3 | XP_006710480.1 | 3 | 361 | |||
| XM_011540910.2 | XP_011539212.1 | 3 | 359 | |||
| XM_017000549.1 | XP_016856038.1 | 3 | 361 | |||
| XM_017000550.1 | XP_016856039.1 | 3 | 361 | |||
| XM_017000551.1 | XP_016856040.1 | 3 | 571 | |||
| XM_017000552.1 | XP_016856041.1 | 3 | 399 | |||
| GenBank mRNA | BC121187.1 | - | 3 | 285 | 3870153 | |
| BC137291.1 | - | 3 | 330 | 9021288 | ||

TMEM201
Genomic Map