siRNA Details
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012758.2 | NP_001012776.1 | 2 | 322 | ||
| XM_011509262.1 | XP_011507564.1 | 2 | 970 | |||
| XM_017000560.1 | XP_016856049.1 | 2 | 970 | |||
| GenBank mRNA | AK091752.1 | - | 1 | 1528 | ||
| AK298083.1 | - | 2 | 323 | |||
| BC046352.1 | - | 1 | 30 | |||
| BG820863.1 | - | 2 | 322 | |||

NUDT17
Genomic Map