siRNA Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168247.2 | NP_001161719.1 | 3 | 818 | ||
| NM_001300859.1 | NP_001287788.1 | 3 | 725 | |||
| NM_144990.3 | NP_659427.3 | 3 | 662 | |||
| XM_005270597.2 | XP_005270654.1 | 2 | 1401 | |||
| XM_005270598.2 | XP_005270655.1 | 2 | 717 | |||
| XM_005270599.2 | XP_005270656.1 | 3 | 725 | |||
| XM_006710432.3 | XP_006710495.1 | 2 | 1812 | |||
| XM_011540943.1 | XP_011539245.1 | 2 | 1348 | |||
| XM_011540945.1 | XP_011539247.1 | 2 | 1285 | |||
| XM_011540947.1 | XP_011539249.1 | 2 | 810 | |||
| XM_011540948.1 | XP_011539250.1 | 3 | 1263 | |||
| XM_011540949.1 | XP_011539251.1 | 2 | 1193 | |||
| XM_011540950.1 | XP_011539252.1 | 2 | 1163 | |||
| XM_011540951.1 | XP_011539253.1 | 3 | 717 | |||
| XM_011540952.2 | XP_011539254.1 | 2 | 1140 | |||
| XM_011540953.2 | XP_011539255.1 | 2 | 1720 | |||
| XM_011540954.1 | XP_011539256.1 | 2 | 1071 | |||
| XM_017000573.1 | XP_016856062.1 | 2 | 1401 | |||
| XM_017000574.1 | XP_016856063.1 | 2 | 1401 | |||
| GenBank mRNA | AK097609.1 | - | 3 | 713 | ||
| AK292296.1 | - | 3 | 788 | |||
| BC022037.2 | - | 3 | 624 | 4826722 | ||
| BC037879.1 | - | 1 | 1474 | 5274032 | ||
| BC050353.1 | - | 1 | 2982 | 5265392 | ||

SLFNL1
Genomic Map