siRNA Details
| Gene Symbol: KRTCAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173852.3 | NP_776251.1 | 2, 3 | 264 | ||
| GenBank mRNA | AK311907.1 | - | 2, 3 | 238 | ||
| AY157577.1 | - | 1, 2 | 149 | |||
| BC029806.1 | - | 2, 3 | 251 | 5189880 | ||
| BC048205.1 | - | 2, 3 | 190 | |||
| BC057233.1 | - | 2, 3 | 273 | 6463372 | ||
| BX647396.1 | - | 2, 3 | 193 | |||

KRTCAP2
Genomic Map