siRNA Details
| Gene Symbol: EPHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005232.4 | NP_005223.4 | 14 | 2309 | ||
| XM_006715880.3 | XP_006715943.1 | 13 | 1937 | |||
| GenBank mRNA | AK290351.1 | - | 14 | 2273 | ||
| AK313263.1 | - | 14 | 2282 | |||
| BC130291.1 | - | 14 | 2293 | 40146322 | ||
| M18391.1 | - | 14 | 2339 | |||
| Z27409.1 | - | 10 | 1367 | |||

EPHA1
Genomic Map