siRNA Details
| Gene Symbol: FAM209A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012971.3 | NP_001012989.2 | 2 | 447 | ||
| GenBank mRNA | BC070179.1 | - | 2 | 409 | ||
| BC107704.1 | - | 2 | 422 | 6614835 | ||
| BX335293.2 | - | 2 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM209A| Gene Symbol: FAM209A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012971.3 | NP_001012989.2 | 2 | 447 | ||
| GenBank mRNA | BC070179.1 | - | 2 | 409 | ||
| BC107704.1 | - | 2 | 422 | 6614835 | ||
| BX335293.2 | - | 2 | 447 | |||