siRNA Details
| Gene Symbol: GKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182536.2 | NP_872342.2 | 4 | 409 | ||
| GenBank mRNA | AF494509.2 | - | 4 | 409 | ||
| AJ966788.1 | - | 4 | 330 | |||
| AY358664.1 | - | 4 | 384 | |||
| AY943908.1 | - | 4 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GKN2| Gene Symbol: GKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182536.2 | NP_872342.2 | 4 | 409 | ||
| GenBank mRNA | AF494509.2 | - | 4 | 409 | ||
| AJ966788.1 | - | 4 | 330 | |||
| AY358664.1 | - | 4 | 384 | |||
| AY943908.1 | - | 4 | 332 | |||