siRNA Details
| Gene Symbol: KCTD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128214.1 | NP_001121686.1 | 3 | 180 | ||
| NM_153331.3 | NP_699162.3 | 2 | 454 | |||
| XM_005264937.2 | XP_005264994.1 | 3 | 1512 | |||
| GenBank mRNA | AK027572.1 | - | 2 | 1046 | ||
| AK074934.1 | - | 2 | 454 | |||
| AK075297.1 | - | 3 | 162 | |||
| BC022893.1 | - | 2 | 196 | 4689885 | ||
| DA359951.1 | - | 3 | 181 | |||

KCTD6
Genomic Map