siRNA Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173546.2 | NP_775817.1 | 5 | 1047 | ||
| XM_005264938.2 | XP_005264995.1 | 5 | 1533 | |||
| XM_005264940.4 | XP_005264997.1 | 5 | 1168 | |||
| XM_006713015.2 | XP_006713078.1 | 5 | 1563 | |||
| XM_006713016.2 | XP_006713079.1 | 5 | 1542 | |||
| GenBank mRNA | AK002164.1 | - | 2 | 382 | ||
| AK294160.1 | - | 5 | 896 | |||
| AY129011.1 | - | 5 | 1004 | |||
| BC013110.2 | - | 1 | 41 | |||
| BC039083.1 | - | 5 | 997 | 5167173 | ||

KLHDC8B
Genomic Map