siRNA Details
| Gene Symbol: TVP23C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135036.1 | NP_001128508.1 | 3 | 379 | ||
| NM_145301.2 | NP_660344.2 | 3 | 379 | |||
| GenBank mRNA | AK310363.1 | - | 3 | 338 | ||
| BC011952.1 | - | 3 | 279 | 3860433 | ||
| Gene Symbol: TVP23C-CDRT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204478.1 | NP_001191407.1 | 3 | 379 | ||
| GenBank mRNA | AB074181.1 | - | 3 | 309 | ||

TVP23C
Genomic Map