siRNA Details
| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288629.1 | NP_001275558.1 | 15 | 2833 | ||
| NM_004440.3 | NP_004431.1 | 15 | 2848 | |||
| XM_005248669.2 | XP_005248726.1 | 15 | 2893 | |||
| XM_017010365.1 | XP_016865854.1 | 15 | 2878 | |||
| GenBank mRNA | AB209269.1 | - | 15 | 2820 | ||
| AK313529.1 | - | 15 | 2820 | |||
| BC126125.1 | - | 15 | 2722 | 8991841 | ||
| BC126151.1 | - | 15 | 2722 | 8991867 | ||
| BC143857.1 | - | 15 | 2707 | 9052373 | ||
| BC143858.1 | - | 15 | 2710 | 9052374 | ||
| L36642.1 | - | 15 | 2792 | |||

EPHA7
Genomic Map