siRNA Details
| Gene Symbol: UNC13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199242.2 | NP_954712.1 | 16 | 1787 | ||
| XM_011524504.1 | XP_011522806.1 | 16 | 1420 | |||
| XM_011524506.1 | XP_011522808.1 | 16 | 1417 | |||
| XM_011524507.2 | XP_011522809.1 | 16 | 1817 | |||
| GenBank mRNA | AJ578444.1 | - | 16 | 1465 | ||
| AK024474.1 | - | 14 | 1689 | |||
| AK300316.1 | - | 17 | 1478 | |||
| AK301529.1 | - | 16 | 1420 | |||
| BC024740.2 | - | 14 | 1216 | |||
| BC067084.1 | - | 16 | 1787 | 5951944 | ||

UNC13D
Genomic Map