siRNA Details
| Gene Symbol: FBXO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142958.1 | NP_001136430.1 | 5 | 677 | ||
| NM_152676.2 | NP_689889.1 | 5 | 705 | |||
| XM_011525858.2 | XP_011524160.1 | 5 | 646 | |||
| XM_011525859.1 | XP_011524161.1 | 5 | 646 | |||
| XM_011525860.1 | XP_011524162.1 | 5 | 646 | |||
| XM_017025603.1 | XP_016881092.1 | 6 | 687 | |||
| XM_017025604.1 | XP_016881093.1 | 5 | 2715 | |||
| GenBank mRNA | AK094215.1 | - | 5 | 617 | ||
| BC029579.1 | - | 5 | 655 | 5268841 | ||

FBXO15
Genomic Map