siRNA Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025300.2 | NP_001020471.2 | 5, 6 | 912 | ||
| XM_006722300.3 | XP_006722363.1 | 5, 6 | 1225 | |||
| GenBank mRNA | AK127824.1 | - | 4, 5 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB12| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025300.2 | NP_001020471.2 | 5, 6 | 912 | ||
| XM_006722300.3 | XP_006722363.1 | 5, 6 | 1225 | |||
| GenBank mRNA | AK127824.1 | - | 4, 5 | 527 | ||