siRNA Details
| Gene Symbol: ZSCAN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152677.2 | NP_689890.1 | 5 | 1748 | ||
| XM_011526607.2 | XP_011524909.1 | 7 | 1929 | |||
| XM_017026458.1 | XP_016881947.1 | 5 | 2148 | |||
| GenBank mRNA | AK092424.1 | - | 5 | 1747 | ||
| BC101738.1 | - | 3 | 1139 | 8069244 | ||
| BC101740.1 | - | 3 | 1139 | 8069246 | ||

ZSCAN4
Genomic Map