siRNA Details
| Gene Symbol: STT3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178862.2 | NP_849193.1 | 7 | 1520 | ||
| XM_011533465.1 | XP_011531767.1 | 7 | 1383 | |||
| XM_017005857.1 | XP_016861346.1 | 7 | 1383 | |||
| XM_017005858.1 | XP_016861347.1 | 7 | 841 | |||
| XM_017005859.1 | XP_016861348.1 | 7 | 914 | |||
| GenBank mRNA | AK074587.1 | - | 7 | 1049 | ||
| AK311488.1 | - | 7 | 1383 | |||
| AX799082.1 | - | 7 | 1022 | |||
| AY074880.1 | - | 7 | 1022 | |||

STT3B
Genomic Map