siRNA Details
| Gene Symbol: STT3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178862.2 | NP_849193.1 | 15 | 2716 | ||
| XM_017005857.1 | XP_016861346.1 | 15 | 2579 | |||
| XM_017005858.1 | XP_016861347.1 | 15 | 2037 | |||
| XM_017005859.1 | XP_016861348.1 | 15 | 2110 | |||
| GenBank mRNA | AK027789.1 | - | 5 | 600 | ||
| AK075380.1 | - | 5 | 691 | |||
| AX799082.1 | - | 15 | 2218 | |||
| AY074880.1 | - | 15 | 2218 | |||
| BC015880.1 | - | 4 | 494 | |||

STT3B
Genomic Map