siRNA Details
| Gene Symbol: SMIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317896.1 | NP_001304825.1 | 5 | 658 | ||
| NM_001317897.1 | NP_001304826.1 | 4 | 367 | |||
| NM_174921.2 | NP_777581.1 | 4 | 513 | |||
| XM_011513640.2 | XP_011511942.1 | 4 | 646 | |||
| XM_017007862.1 | XP_016863351.1 | 5 | 562 | |||
| XM_017007863.1 | XP_016863352.1 | 4 | 403 | |||
| GenBank mRNA | AK023351.1 | - | 4 | 376 | ||
| BC008502.1 | - | 4 | 376 | 4295121 | ||
| BQ049060.1 | - | 4 | 358 | |||
| HY155982.1 | - | 4 | 514 | |||

SMIM14
Genomic Map