siRNA Details
| Gene Symbol: RWDD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307922.1 | NP_001294851.1 | 3 | 413 | ||
| NM_152682.3 | NP_689895.2 | 3 | 417 | |||
| GenBank mRNA | AK021702.1 | - | 3 | 340 | ||
| AK315515.1 | - | 3 | 351 | |||
| AL540937.3 | - | 3 | 354 | |||
| AL559547.3 | - | 3 | 350 | |||
| BC017472.1 | - | 3 | 352 | 3909581 | ||
| BC067752.1 | - | 3 | 376 | 5270461 | ||
| BC107432.1 | - | 3 | 198 | 6104330 | ||
| DA250301.1 | - | 3 | 417 | |||
| DA766837.1 | - | 3 | 416 | |||

RWDD4
Genomic Map