siRNA Details
| Gene Symbol: GAPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304428.1 | NP_001291357.1 | 3 | 949 | ||
| NM_001304429.1 | NP_001291358.1 | 3 | 942 | |||
| NM_001304431.1 | NP_001291360.1 | 3 | 850 | |||
| NM_152687.3 | NP_689900.1 | 3 | 787 | |||
| GenBank mRNA | AK090960.1 | - | 3 | 721 | ||
| BC063534.1 | - | 1 | 479 | 4430075 | ||

GAPT
Genomic Map