siRNA Details
| Gene Symbol: CMYA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153610.4 | NP_705838.3 | 2 | 2740 | ||
| XM_017009212.1 | XP_016864701.1 | 2 | 2608 | |||
| GenBank mRNA | AL832347.1 | - | 2 | 2542 | ||
| AL832376.1 | - | 2 | 2699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CMYA5| Gene Symbol: CMYA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153610.4 | NP_705838.3 | 2 | 2740 | ||
| XM_017009212.1 | XP_016864701.1 | 2 | 2608 | |||
| GenBank mRNA | AL832347.1 | - | 2 | 2542 | ||
| AL832376.1 | - | 2 | 2699 | |||