siRNA Details
Gene Symbol: OSTCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028496.1 | 3 | 1005 | |||
GenBank mRNA | BC024224.1 | - | 3 | 1009 | 3998034 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OSTCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028496.1 | 3 | 1005 | |||
GenBank mRNA | BC024224.1 | - | 3 | 1009 | 3998034 |