siRNA Details
| Gene Symbol: TMEM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097620.1 | NP_001091089.1 | 9 | 2253 | ||
| XM_006715653.1 | XP_006715716.1 | 10 | 2311 | |||
| XM_006715655.2 | XP_006715718.1 | 8 | 3245 | |||
| XM_011515164.1 | XP_011513466.1 | 10 | 2251 | |||
| XM_017011815.1 | XP_016867304.1 | 8 | 3245 | |||
| GenBank mRNA | AK075043.1 | - | 9 | 2114 | ||
| AK096790.1 | - | 6 | 2814 | |||
| AK126312.1 | - | 8 | 2053 | |||
| AK172850.1 | - | 9 | 2029 | |||
| BC026694.1 | - | 9 | 2031 | 3850351 | ||

TMEM184A
Genomic Map