siRNA Details
| Gene Symbol: TMEM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097620.1 | NP_001091089.1 | 9 | 3997 | ||
| XM_017011815.1 | XP_016867304.1 | 8 | 4989 | |||
| GenBank mRNA | AK126312.1 | - | 8 | 3797 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM184A| Gene Symbol: TMEM184A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097620.1 | NP_001091089.1 | 9 | 3997 | ||
| XM_017011815.1 | XP_016867304.1 | 8 | 4989 | |||
| GenBank mRNA | AK126312.1 | - | 8 | 3797 | ||