siRNA Details
| Gene Symbol: C8orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040032.1 | NP_001035121.1 | 2 | 238 | ||
| XM_011543816.2 | XP_011542118.1 | 2 | 339 | |||
| GenBank mRNA | AK131443.1 | - | 2 | 222 | ||
| BC065837.1 | - | 2 | 238 | |||
| BC132852.1 | - | 2 | 238 | 40146874 | ||
| BC132854.1 | - | 2 | 238 | 40146876 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026359, AK005778, AK016302, BC048583.

C8orf74
Genomic Map