siRNA Details
| Gene Symbol: HTRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153692.3 | NP_710159.1 | 9 | 1410 | ||
| GenBank mRNA | AK075205.1 | - | 9 | 1381 | ||
| AK300197.1 | - | 9 | 1289 | |||
| BC057765.1 | - | 9 | 1342 | 30348811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTRA4| Gene Symbol: HTRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153692.3 | NP_710159.1 | 9 | 1410 | ||
| GenBank mRNA | AK075205.1 | - | 9 | 1381 | ||
| AK300197.1 | - | 9 | 1289 | |||
| BC057765.1 | - | 9 | 1342 | 30348811 | ||