siRNA Details
| Gene Symbol: CT83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017978.3 | NP_001017978.1 | 2 | 420 | ||
| GenBank mRNA | AK000618.1 | - | 2 | 390 | ||
| AK026566.1 | - | 2 | 390 | |||
| BC062223.1 | - | 2 | 367 | 6101827 | ||
| HY170569.1 | - | 2 | 421 | |||

CT83
Genomic Map