siRNA Details
| Gene Symbol: ZCCHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152694.2 | NP_689907.1 | 2 | 1489 | ||
| GenBank mRNA | AK096184.1 | - | 2 | 1489 | ||
| BC136548.1 | - | 1 | 1397 | 9020538 | ||
| BC136549.1 | - | 1 | 1397 | 9020539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZCCHC5| Gene Symbol: ZCCHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152694.2 | NP_689907.1 | 2 | 1489 | ||
| GenBank mRNA | AK096184.1 | - | 2 | 1489 | ||
| BC136548.1 | - | 1 | 1397 | 9020538 | ||
| BC136549.1 | - | 1 | 1397 | 9020539 | ||