siRNA Details
| Gene Symbol: CERS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290341.2 | NP_001277270.1 | 11 | 1394 | ||
| NM_001290342.2 | NP_001277271.1 | 10 | 1220 | |||
| NM_001290343.1 | NP_001277272.1 | 10 | 1131 | |||
| NM_178842.4 | NP_849164.2 | 10 | 1127 | |||
| XM_011521355.2 | XP_011519657.1 | 11 | 1437 | |||
| XM_011521357.2 | XP_011519659.1 | 11 | 1331 | |||
| XM_011521358.1 | XP_011519660.1 | 11 | 1215 | |||
| XM_017022002.1 | XP_016877491.1 | 10 | 1211 | |||
| XM_017022003.1 | XP_016877492.1 | 10 | 1136 | |||
| XM_017022004.1 | XP_016877493.1 | 9 | 1047 | |||
| GenBank mRNA | BC028703.1 | - | 10 | 1208 | 4837657 | |
| BC034500.1 | - | 10 | 1083 | 4838423 | ||
| BC034970.1 | - | 10 | 1129 | 4822006 | ||

CERS3
Genomic Map