siRNA Details
| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 3 | 322 | ||
| GenBank mRNA | AJ293620.1 | - | 3 | 282 | ||
| AY358849.1 | - | 3 | 284 | |||
| BC019696.2 | - | 3 | 275 | |||
| BC032340.1 | - | 3 | 286 | 5192975 | ||

DHRSX
Genomic Map