siRNA Details
| Gene Symbol: AMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152704.3 | NP_689917.2 | 1 | 2054 | ||
| NM_199138.2 | NP_954589.1 | 3 | 1388 | |||
| XM_005266279.3 | XP_005266336.1 | 2 | 2034 | |||
| XM_005266280.3 | XP_005266337.1 | 2 | 1944 | |||
| XM_017020415.1 | XP_016875904.1 | 2 | 1708 | |||
| GenBank mRNA | AK055049.1 | - | 3 | 1388 | ||
| AK098343.1 | - | 1 | 1254 | |||
| AK098796.1 | - | 2 | 329 | |||
| AL832627.1 | - | 1 | 107 | |||
| BC032653.1 | - | 2 | 974 | |||
| BC041392.1 | - | 1 | 2062 | 5277030 | ||

AMER2
Genomic Map