siRNA Details
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 12 | 2966 | ||
GenBank mRNA | AK094502.1 | - | 2 | 924 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ATAD3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039211.2 | NP_001034300.2 | 12 | 2966 | ||
GenBank mRNA | AK094502.1 | - | 2 | 924 |