siRNA Details
| Gene Symbol: MTIF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166261.1 | NP_001159733.1 | 3 | 179 | ||
| NM_001166262.1 | NP_001159734.1 | 3 | 249 | |||
| NM_001166263.1 | NP_001159735.1 | 4 | 278 | |||
| NM_152912.4 | NP_690876.3 | 3 | 217 | |||
| XM_006719771.3 | XP_006719834.1 | 4 | 265 | |||
| XM_006719772.3 | XP_006719835.1 | 3 | 496 | |||
| XM_011534957.2 | XP_011533259.1 | 4 | 739 | |||
| XM_011534958.2 | XP_011533260.1 | 4 | 523 | |||
| XM_011534959.2 | XP_011533261.1 | 4 | 688 | |||
| XM_011534960.2 | XP_011533262.1 | 4 | 353 | |||
| XM_011534961.2 | XP_011533263.1 | 4 | 941 | |||
| XM_011534962.2 | XP_011533264.1 | 4 | 518 | |||
| XM_011534963.2 | XP_011533265.1 | 4 | 1106 | |||
| XM_017020417.1 | XP_016875906.1 | 3 | 123 | |||
| GenBank mRNA | AF410851.1 | - | 2 | 84 | ||
| AK094420.1 | - | 1 | 1278 | |||
| BC039599.1 | - | 3 | 170 | 3855096 | ||
| BC046166.1 | - | 3 | 189 | 3460180 | ||
| BG617536.1 | - | 3 | 218 | |||
| BI460170.1 | - | 4 | 272 | |||
| BM548061.1 | - | 2 | 142 | |||
| CN272769.1 | - | 4 | 202 | |||
| DB456678.1 | - | 3 | 251 | |||
| DB470026.1 | - | 3 | 179 | |||

MTIF3
Genomic Map