siRNA Details
| Gene Symbol: GSX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145657.2 | NP_663632.1 | 2 | 586 | ||
| GenBank mRNA | AB044157.1 | - | 2 | 586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSX1| Gene Symbol: GSX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145657.2 | NP_663632.1 | 2 | 586 | ||
| GenBank mRNA | AB044157.1 | - | 2 | 586 | ||