siRNA Details
| Gene Symbol: OR4C11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004700.2 | NP_001004700.2 | 1 | 593 | ||
| GenBank mRNA | BC140727.1 | - | 1 | 593 | 9021595 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C11| Gene Symbol: OR4C11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004700.2 | NP_001004700.2 | 1 | 593 | ||
| GenBank mRNA | BC140727.1 | - | 1 | 593 | 9021595 | |