siRNA Details
| Gene Symbol: OR8H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005199.1 | NP_001005199.1 | 1 | 629 | ||
| GenBank mRNA | BC136909.1 | - | 1 | 657 | 9020900 | |
| BC136910.1 | - | 1 | 657 | 9020901 | ||
| Gene Symbol: OR8H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005201.1 | NP_001005201.1 | 1 | 632 | ||

OR8H1
Genomic Map